4,5-Dibromo-1H-imidazole 98%

Supplier: Ambeed
Danger

A852970-100MG A852970-1G A852970-250MG
77118-546EA 36.05 USD
77118-546 77118-548 77118-550
4,5-Dibromo-1H-imidazole 98%
4,5-Dibromo-1H-imidazole

Precaución: for Research Use Only

Formula: C₃H₂Br₂N₂
MDL Number: MFCD00091826
CAS Number: 2302-30-9
UN: 2928
ADR: 6.1,8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR