4-Aminobenzamidoxime 97%

Supplier: Ambeed
Danger

Synonyms: 4-Aminobenzamide oxime, 4-Amino-N'-hydroxybenzimidamide

A848084-1G A848084-250MG A848084-25G A848084-5G
77118-538EA 41.2 USD
77118-538 77118-540 77118-542 77118-544
4-Aminobenzamidoxime 97%
4-Aminobenzamidoxime

Precaución: for Research Use Only

Formula: C₇H₉N₃O
MW: 151.17
MDL Number: MFCD08061151
CAS Number: 277319-62-7
UN: 2811
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR