8-Fluoro-3-iodoquinoline 97%

Supplier: Ambeed
Warning

A847594-100MG A847594-1G A847594-250MG
77118-532EA 51.99 USD
77118-532 77118-534 77118-536
8-Fluoro-3-iodoquinoline 97%
8-Fluoro-3-iodoquinoline

Precaución: for Research Use Only

Formula: C₉H₅FIN
MW: 273.046 g/mol
MDL Number: MFCD07700291
CAS Number: 866782-59-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR