3-[2-(2-Ethoxycarbonyl-ethoxy)-ethoxy]-propionic acid 97%

Supplier: Ambeed
Warning

Synonyms: 3-(2-(3-Ethoxy-3-oxopropoxy)ethoxy)propanoic acid, Acid-PEG2-ethyl propionate

A846940-250MG A846940-5G A846940-100MG A846940-1G
77118-528EA 33.99 USD
77118-528 77118-530 77686-880 77686-881
3-[2-(2-Ethoxycarbonyl-ethoxy)-ethoxy]-propionic acid 97%
3-[2-(2-Ethoxycarbonyl-ethoxy)-ethoxy]-propionic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₈O₆
MDL Number: MFCD04115349
CAS Number: 886362-90-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR