tert-Butyl 4-(benzo[d]thiazol-2-yl)piperidine-1-carboxylate 98%

Supplier: Ambeed
Warning

A846252-100MG A846252-1G A846252-250MG
77118-518EA 16.98 USD
77118-518 77118-520 77118-522
tert-Butyl 4-(benzo[d]thiazol-2-yl)piperidine-1-carboxylate 98%
tert-Butyl 4-(benzo[d]thiazol-2-yl)piperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₇H₂₂N₂O₂S
MW: 318.43 g/mol
MDL Number: MFCD21105859
CAS Number: 1373879-28-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR