4-Chloro-8-methoxy-2-methylquinoline 98%

Supplier: Ambeed
Danger

A845456-1G A845456-250MG A845456-25G A845456-5G
77118-510EA 9.27 USD
77118-510 77118-512 77118-514 77118-516
4-Chloro-8-methoxy-2-methylquinoline 98%
4-Chloro-8-methoxy-2-methylquinoline

Precaución: for Research Use Only

Formula: C₁₁H₁₀ClNO
MDL Number: MFCD00272401
CAS Number: 64951-58-2
UN: 2923
ADR: 8,6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR