2-Amino-3-bromo-5-nitrobenzonitrile 98%

Supplier: Ambeed
Warning

A840924-100G A840924-25G A840924-5G
77118-478EA 82.76 USD
77118-478 77118-480 77118-482
2-Amino-3-bromo-5-nitrobenzonitrile 98%
2-Amino-3-bromo-5-nitrobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₄BrN₃O₂
MW: 242.03 g/mol
MDL Number: MFCD00054185
CAS Number: 17601-94-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR