Ethyl-m-toluate 99%

Supplier: Ambeed
Warning

A840795-1G A840795-25G A840795-5G
77118-472EA 5.15 USD
77118-472 77118-474 77118-476
Ethyl-m-toluate 99%
Ethyl-m-toluate

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₂
MDL Number: MFCD00009114
CAS Number: 120-33-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR