2-Fluoro-3-nitro-5-picoline 98%

Supplier: Ambeed
Warning

Synonyms: 2-Fluoro-5-methyl-3-nitropyridine, 2-Fluoro-3-nitro-5-methylpyridine

A839478-10G A839478-1G
77118-468EA 93.73 USD
77118-468 77118-470
2-Fluoro-3-nitro-5-picoline 98%
2-Fluoro-3-nitro-5-picoline

Precaución: for Research Use Only

Formula: C₆H₅FN₂O₂
MW: 156.12
MDL Number: MFCD09475421
CAS Number: 19346-44-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR