α-Bromo-3-chloro-2-fluorotoluene 97%

Supplier: Ambeed
Danger

Synonyms: 3-Chloro-2-fluorobenzyl bromide, 1-(Bromomethyl)-3-chloro-2-fluorobenzene

A838225-100G A838225-10G A838225-1G A838225-25G A838225-5G
77118-452EA 98.87 USD
77118-452 77118-454 77118-456 77118-458 77118-460
α-Bromo-3-chloro-2-fluorotoluene 97%
α-Bromo-3-chloro-2-fluorotoluene

Precaución: for Research Use Only

Formula: C₇H₅BrClF
MDL Number: MFCD00042503
CAS Number: 85070-47-9
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR