(R)-5,5'-Bis[di(3,5-di-tert-butyl-4-methoxyphenyl)phosphino]-4,4'-bi-1,3-benzodioxole ≥98%, ee 99%
Supplier: Ambeed
|
Warning
|
A836153-100MG
A836153-1G
A836153-250MG
A836153-25G
A836153-5G
77118-442EA
17.52
USD
77118-442
77118-444
77118-446
77118-448
77118-450
(R)-5,5'-Bis[di(3,5-di-tert-butyl-4-methoxyphenyl)phosphino]-4,4'-bi-1,3-benzodioxole ≥98%, ee 99%
(R)-5,5'-Bis[di(3,5-di-tert-butyl-4-methoxyphenyl)phosphino]-4,4'-bi-1,3-benzodioxole
Precaución: for Research Use Only
Formula:
C₇₄H₁₀₀O₈P₂ |
MDL Number:
MFCD09753003 CAS Number: 566940-03-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...