Dihydrotanshinone I 98%

Supplier: Ambeed
Danger

A831392-100MG A831392-250MG
77118-432EA 223.51 USD
77118-432 77118-434
Dihydrotanshinone I 98%
Dihydrotanshinone I

Precaución: for Research Use Only

Formula: C₁₈H₁₄O₃
MDL Number: MFCD28016070
CAS Number: 87205-99-0
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR