4-Bromo-2-iodobenzonitrile 95%

Supplier: Ambeed
Warning

A823564-250MG A823564-25G A823564-5G
77118-424EA 35.02 USD
77118-424 77118-426 77573-628
4-Bromo-2-iodobenzonitrile 95%
4-Bromo-2-iodobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₃BrIN
MDL Number: MFCD09907887
CAS Number: 944276-67-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR