2,4-Dimethylanisole 98%

Supplier: Ambeed
Danger

A819713-1G A819713-25G A819713-5G
77118-418EA 11.33 USD
77118-418 77118-420 77118-422
2,4-Dimethylanisole 98%
2,4-Dimethylanisole

Precaución: for Research Use Only

Formula: C₉H₁₂O
MDL Number: MFCD00008377
CAS Number: 6738-23-4
UN: 1760
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR