4-Chloro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Chloro-5-formylphenylboronic acid pinacol ester
A819328-100MG
A819328-1G
A819328-250MG
77118-412EA
16.98
USD
77118-412
77118-414
77118-416
4-Chloro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde 95%
4-Chloro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde
Precaución: for Research Use Only
Formula:
C₁₃H₁₆BClO₃ MW: 266.52 g/mol |
MDL Number:
MFCD18731013 CAS Number: 1112209-14-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...