2-Amino-3,5-dichlorobenzonitrile 98%

Supplier: Ambeed
Warning

A816819-1G A816819-25G A816819-5G
77118-410EA 7.43 USD
77118-410 77686-842 77686-843
2-Amino-3,5-dichlorobenzonitrile 98%
2-Amino-3,5-dichlorobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₄Cl₂N₂
MW: 187.03 g/mol
MDL Number: MFCD00158947
CAS Number: 36764-94-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR