4'-Methoxybiphenyl-4-carboxylic acid 98%

Supplier: Ambeed
Warning

A814482-1G A814482-25G A814482-5G
77118-396EA 25.76 USD
77118-396 77118-398 77118-400
4'-Methoxybiphenyl-4-carboxylic acid 98%
4'-Methoxybiphenyl-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₂O₃
MDL Number: MFCD00185792
CAS Number: 725-14-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR