4-(4-Fluoro-3-(4-methoxypiperidine-1-carbonyl)benzyl)phthalazin-1(2H)-one 98+%

Supplier: Ambeed
Warning

A810897-10MG A810897-1MG A810897-25MG A810897-50MG A810897-5MG
77118-382EA 55.62 USD
77118-382 77118-384 77118-386 77118-388 77118-390
4-(4-Fluoro-3-(4-methoxypiperidine-1-carbonyl)benzyl)phthalazin-1(2H)-one 98+%
4-(4-Fluoro-3-(4-methoxypiperidine-1-carbonyl)benzyl)phthalazin-1(2H)-one

Precaución: for Research Use Only

Formula: C₂₂H₂₂FN₃O₃
MDL Number: MFCD24386811
CAS Number: 1174043-16-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR