2-Ethynyl-5-fluoropyridine 98%

Supplier: Ambeed
Warning

A809773-250MG A809773-1G
77118-374EA 17.52 USD
77118-374 77686-840
2-Ethynyl-5-fluoropyridine 98%
2-Ethynyl-5-fluoropyridine

Precaución: for Research Use Only

Formula: C₇H₄FN
MDL Number: MFCD03095259
CAS Number: 884494-34-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR