5-Chloro-2-hydroxyphenylboronic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-Chloro-2-hydroxybenzeneboronic acid
A807359-25G
A807359-1G
77118-372EA
504.99
USD
77118-372
77686-837
5-Chloro-2-hydroxyphenylboronic acid 98%
5-Chloro-2-hydroxyphenylboronic acid
Precaución: for Research Use Only
Formula:
C₆H₆BClO₃ MW: 172.37 g/mol |
MDL Number:
MFCD06659863 CAS Number: 89488-25-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...