4-Bromobenzo[d]thiazol-5-amine 98%

Supplier: Ambeed
Warning

A797315-1G A797315-250MG A797315-5G
77118-360EA 231.28 USD
77118-360 77118-362 77118-364
4-Bromobenzo[d]thiazol-5-amine 98%
4-Bromobenzo[d]thiazol-5-amine

Precaución: for Research Use Only

Formula: C₇H₅BrN₂S
MW: 229.10 g/mol
MDL Number: MFCD05865268
CAS Number: 769-19-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR