3-Methylglutaric acid 98%

Supplier: Ambeed
Warning

A791966-100G A791966-1G A791966-5G
77118-344EA 177.18 USD
77118-344 77118-346 77118-348
3-Methylglutaric acid 98%
3-Methylglutaric acid

Precaución: for Research Use Only

Formula: C₆H₁₀O₄
MW: 146.1412 g/mol
MDL Number: MFCD00002727
CAS Number: 626-51-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR