Methyl acetopyruvate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 2,4-dioxopentanoate

A791218-100G A791218-1G A791218-25G A791218-5G
77118-336EA 353.28 USD
77118-336 77118-338 77118-340 77118-342
Methyl acetopyruvate 98%
Methyl acetopyruvate

Precaución: for Research Use Only

Formula: C₆H₈O₄
MW: 144.13 g/mol
MDL Number: MFCD00043638
CAS Number: 20577-61-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR