4-(Trifluoromethyl)nicotinaldehyde 95%

Supplier: Ambeed
Danger

A791113-1G A791113-5G
77118-332EA 83.43 USD
77118-332 77118-334
4-(Trifluoromethyl)nicotinaldehyde 95%
4-(Trifluoromethyl)nicotinaldehyde

Precaución: for Research Use Only

Formula: C₇H₄F₃NO
MW: 175.11 g/mol
MDL Number: MFCD07781251
CAS Number: 1083197-78-7
UN: 2810
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR