(+/-)-Phenyl(pyridin-2-yl)methanol 98%

Supplier: Ambeed
Warning

A783793-1G A783793-5G
77118-296EA 52.53 USD
77118-296 77118-298
(+/-)-Phenyl(pyridin-2-yl)methanol 98%
(+/-)-Phenyl(pyridin-2-yl)methanol

Precaución: for Research Use Only

Formula: C₁₂H₁₁NO
MDL Number: MFCD00186514
CAS Number: 14159-57-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR