4,6-Dichloropicolinamide 95%

Supplier: Ambeed
Warning

A781963-100MG A781963-250MG
77118-292EA 85.94 USD
77118-292 77118-294
4,6-Dichloropicolinamide 95%
4,6-Dichloropicolinamide

Precaución: for Research Use Only

Formula: C₆H₄Cl₂N₂O
MW: 191.01 g/mol
MDL Number: MFCD16038703
CAS Number: 98141-39-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR