Trimagnesium bis((R)-5-((S)-2-hydroxy-1-oxidoethyl)-4-oxido-2-oxo-2,5-dihydrofuran-3-yl phosphate) 98%

Supplier: Ambeed
Warning

A757954-100G A757954-25G
77118-210EA 49.87 USD
77118-210 77118-212
Trimagnesium bis((R)-5-((S)-2-hydroxy-1-oxidoethyl)-4-oxido-2-oxo-2,5-dihydrofuran-3-yl phosphate) 98%
Trimagnesium bis((R)-5-((S)-2-hydroxy-1-oxidoethyl)-4-oxido-2-oxo-2,5-dihydrofuran-3-yl phosphate)

Precaución: for Research Use Only

Formula: C₁₂H₁₀Mg₃O₁₈P₂
MW: 577.06 g/mol
MDL Number: MFCD11045932
CAS Number: 113170-55-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR