2-(1-((Benzyloxy)carbonyl)pyrrolidin-3-yl)acetic acid 96%

Supplier: Ambeed
Warning

A756993-100MG A756993-1G A756993-250MG A756993-5G
77118-194EA 51.99 USD
77118-194 77118-196 77118-198 77118-200
2-(1-((Benzyloxy)carbonyl)pyrrolidin-3-yl)acetic acid 96%
2-(1-((Benzyloxy)carbonyl)pyrrolidin-3-yl)acetic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₇NO₄
MW: 263.28 g/mol
MDL Number: MFCD02179025
CAS Number: 886362-65-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR