(S)-2-Methylmorpholine 97%

Supplier: Ambeed
Danger

A754936-1G A754936-250MG A754936-5G A754936-100MG
77118-178EA 117.76 USD
77118-178 77118-180 77118-182 77686-763
(S)-2-Methylmorpholine 97%
(S)-2-Methylmorpholine

Precaución: for Research Use Only

Formula: C₅H₁₁NO
MW: 101.1469 g/mol
MDL Number: MFCD11506701
CAS Number: 74572-13-7
UN: 2734
ADR: 8,3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR