α-Chloro-2,6-difluorotoulene 98%

Supplier: Ambeed
Danger

A752620-5G A752620-1G
77118-176EA 11.33 USD
77118-176 77686-760
α-Chloro-2,6-difluorotoulene 98%
α-Chloro-2,6-difluorotoulene

Precaución: for Research Use Only

Formula: C₇H₅ClF₂
MDL Number: MFCD00191346
CAS Number: 697-73-4
UN: 1759
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR