3,4-Difluorobenzonitrile 98%

Supplier: Ambeed
Warning

A751795-100G A751795-10G A751795-25G A751795-500G A751795-5G
77118-160EA 37.14 USD
77118-160 77118-162 77118-164 77118-166 77118-168
3,4-Difluorobenzonitrile 98%
3,4-Difluorobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₃F₂N
MW: 139.1 g/mol
MDL Number: MFCD00011666
CAS Number: 64248-62-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR