2-(4-(2-(2-Methoxyethoxy)ethoxy)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95+%

Supplier: Ambeed
Warning

A751690-100MG A751690-1G A751690-250MG
77118-154EA 37.14 USD
77118-154 77118-156 77118-158
2-(4-(2-(2-Methoxyethoxy)ethoxy)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95+%
2-(4-(2-(2-Methoxyethoxy)ethoxy)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₇H₂₇BO₅
MW: 322.20 g/mol
MDL Number: MFCD27996253
CAS Number: 1366602-62-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR