Ethyl tetrahydropyran-4-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl tetrahydro-2H-pyran-4-carboxylate

A743486-10G A743486-1G A743486-5G
77118-140EA 122.01 USD
77118-140 77118-142 77118-144
Ethyl tetrahydropyran-4-carboxylate 98%
Ethyl tetrahydropyran-4-carboxylate

Precaución: for Research Use Only

Formula: C₈H₁₄O₃
MW: 158.2 g/mol
MDL Number: MFCD08702763
CAS Number: 96835-17-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR