3-Formyl-1H-indole-2-carboxylic acid 95+%

Supplier: Ambeed
Warning

A742731-100MG A742731-1G A742731-250MG
77118-134EA 7.21 USD
77118-134 77118-136 77118-138
3-Formyl-1H-indole-2-carboxylic acid 95+%
3-Formyl-1H-indole-2-carboxylic acid
Formula: C₁₀H₇NO₃
MDL Number: MFCD06809647
CAS Number: 28737-34-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR