2-Amino-3,5-dibromo-6-methylpyridine 98%
Supplier: Ambeed
|
Warning
|
A741853-100G
A741853-10G
A741853-1G
A741853-25G
77118-122EA
94.76
USD
77118-122
77118-124
77118-126
77118-128
2-Amino-3,5-dibromo-6-methylpyridine 98%
2-Amino-3,5-dibromo-6-methylpyridine
Precaución: for Research Use Only
Formula:
C₆H₆Br₂N₂ |
MDL Number:
MFCD00068229 CAS Number: 91872-10-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...