1-(2-Bromophenyl)piperidine 97%

Supplier: Ambeed
Warning

A737693-1G A737693-5G
77118-112EA 9.26 USD
77118-112 77118-114
1-(2-Bromophenyl)piperidine 97%
1-(2-Bromophenyl)piperidine

Precaución: for Research Use Only

Formula: C₁₁H₁₄BrN
MDL Number: MFCD04112586
CAS Number: 156808-79-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR