4-Chloro-2-(ethylthio)-6-methylpyrimidine 95%

Supplier: Ambeed
Warning

A728634-100MG A728634-1G A728634-250MG
77118-090EA 40.32 USD
77118-090 77118-092 77118-094
4-Chloro-2-(ethylthio)-6-methylpyrimidine 95%
4-Chloro-2-(ethylthio)-6-methylpyrimidine

Precaución: for Research Use Only

Formula: C₇H₉ClN₂S
MW: 188.68 g/mol
MDL Number: MFCD00023245
CAS Number: 22727-18-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR