Aminoacetaldehyde dimethyl acetal 98%

Supplier: Ambeed
Danger

Synonyms: 2,2-Dimethoxyethanamine

A727306-10G A727306-25G A727306-5G
77118-066EA 6.37 USD
77118-066 77118-068 77118-070
Aminoacetaldehyde dimethyl acetal 98%
Aminoacetaldehyde dimethyl acetal

Precaución: for Research Use Only

Formula: C₄H₁₁NO₂
MW: 105.14 g/mol
MDL Number: MFCD00008135
CAS Number: 22483-09-6
UN: 2924
ADR: 3,8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR