2-Heptyl-1H-benzo[d]imidazole 95%

Supplier: Ambeed
Warning

A722975-1G A722975-25G
77118-048EA 10.61 USD
77118-048 77118-050
2-Heptyl-1H-benzo[d]imidazole 95%
2-Heptyl-1H-benzo[d]imidazole

Precaución: for Research Use Only

Formula: C₁₄H₂₀N₂
MW: 216.32 g/mol
MDL Number: MFCD00459384
CAS Number: 5851-49-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR