1-(6,8-Difluoro-2-methylquinolin-4-yl)-3-(4-(dimethylamino)phenyl)urea 98%

Supplier: Ambeed
Warning

A714872-1MG A714872-5MG
77118-020EA 16.48 USD
77118-020 77118-022
1-(6,8-Difluoro-2-methylquinolin-4-yl)-3-(4-(dimethylamino)phenyl)urea 98%
1-(6,8-Difluoro-2-methylquinolin-4-yl)-3-(4-(dimethylamino)phenyl)urea

Precaución: for Research Use Only

Formula: C₁₉H₁₈F₂N₄O
MDL Number: MFCD06411575
CAS Number: 288150-92-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR