2-Bromo-5-fluorobenzylamine 98%

Supplier: Ambeed
Warning

A714060-100G A714060-1G A714060-25G A714060-5G
77118-012EA 311.91 USD
77118-012 77118-014 77118-016 77118-018
2-Bromo-5-fluorobenzylamine 98%
2-Bromo-5-fluorobenzylamine

Precaución: for Research Use Only

Formula: C₇H₇BrFN
MW: 204.03 g/mol
MDL Number: MFCD06212850
CAS Number: 747392-34-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR