Ethyl 1-Boc-3-methylpiperidine-3-carboxylate 95%
Supplier: Ambeed
|
Warning
|
A714024-1G
A714024-250MG
77118-008EA
100.79
USD
77118-008
77118-010
Ethyl 1-Boc-3-methylpiperidine-3-carboxylate 95%
Ethyl 1-Boc-3-methylpiperidine-3-carboxylate
Precaución: for Research Use Only
Formula:
C₁₄H₂₅NO₄ MW: 271.35 g/mol |
MDL Number:
MFCD11559117 CAS Number: 278789-43-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...