α,α,α,3-Tetrafluorotoluene 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
3-Fluorobenzotrifluoride, 1-Fluoro-3-(trifluoromethyl)benzene
A713473-10G
A713473-25G
77118-004EA
5.31
USD
77118-004
77118-006
α,α,α,3-Tetrafluorotoluene 98%
α,α,α,3-Tetrafluorotoluene
Precaución: for Research Use Only
Formula:
C₇H₄F₄ MW: 164.1 g/mol |
MDL Number:
MFCD00000382 CAS Number: 401-80-9 UN: 1993 ADR: 3,Ⅱ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...