α-Bromo-2,5-dichlorotoluene 98%

Supplier: Ambeed
Danger

Synonyms: 2,5-Dichlorobenzyl bromide, 2-(Bromomethyl)-1,4-dichlorobenzene

A710824-10G A710824-1G A710824-25G A710824-5G
77117-988EA 51.5 USD
77117-988 77117-990 77117-992 77117-994
α-Bromo-2,5-dichlorotoluene 98%
α-Bromo-2,5-dichlorotoluene

Precaución: for Research Use Only

Formula: C₇H₅BrCl₂
MW: 239.93 g/mol
Boiling Pt: 259
MDL Number: MFCD00236034
CAS Number: 85482-13-9
UN: 3261
ADR: 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR