4-Bromo-1H-benzo[d]imidazol-5-amine 95%
Supplier: Ambeed
|
Warning
|
A709948-100MG
A709948-250MG
77117-972EA
21.63
USD
77117-972
77117-974
4-Bromo-1H-benzo[d]imidazol-5-amine 95%
4-Bromo-1H-benzo[d]imidazol-5-amine
Precaución: for Research Use Only
Formula:
C₇H₆BrN₃ |
MDL Number:
MFCD03426349 CAS Number: 177843-26-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...