Ethyl-2,4,5-trifluorobenzoate 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2,4,5-trifluorobenzoate

A708732-100G A708732-1G A708732-25G A708732-5G
77117-964EA 152.77 USD
77117-964 77117-966 77117-968 77117-970
Ethyl-2,4,5-trifluorobenzoate 95%
Ethyl 2,4,5-trifluorobenzoate

Precaución: for Research Use Only

Formula: C₉H₇F₃O₂
MW: 204.1459 g/mol
MDL Number: MFCD03094444
CAS Number: 351354-41-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR