3-Amino-5-(methoxycarbonyl)benzoic acid 97%

Supplier: Ambeed
Warning

A705954-250MG A705954-5G A705954-1G
77117-942EA 10.31 USD
77117-942 77686-692 77686-691
3-Amino-5-(methoxycarbonyl)benzoic acid 97%
3-Amino-5-(methoxycarbonyl)benzoic acid

Precaución: for Research Use Only

Formula: C9H9NO4
MDL Number: MFCD00224870
CAS Number: 28179-47-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR