1-Methyl-4-nitro-1H-imidazol-5-amine 95%

Supplier: Ambeed
Warning

Synonyms: Azathioprine Related Compound A

A702114-1G A702114-250MG
77117-926EA 165.51 USD
77117-926 77117-928
1-Methyl-4-nitro-1H-imidazol-5-amine 95%
1-Methyl-4-nitro-1H-imidazol-5-amine

Precaución: for Research Use Only

Formula: C₄H₆N₄O₂
MW: 142.11 g/mol
MDL Number: MFCD00516996
CAS Number: 4531-54-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR