α-Bromo-4-chlorotoulene 98%

Supplier: Ambeed
Danger

Synonyms: alpha-Bromo-4-chlorotoluene, 1-(Bromomethyl)-4-chlorobenzene, 4-Chlorobenzyl bromide

A698850-100G A698850-10G A698850-1G A698850-25G A698850-5G
77117-910EA 93.36 USD
77117-910 77117-912 77117-914 77117-916 77117-918
α-Bromo-4-chlorotoulene 98%
α-Bromo-4-chlorotoulene

Precaución: for Research Use Only

Formula: C₇H₆BrCl
MW: 205.48 g/mol
MDL Number: MFCD00040714
CAS Number: 622-95-7
UN: 1759
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR