2-(Cyclopropylmethoxy)isonicotinonitrile 95%

Supplier: Ambeed
Warning

A698783-100MG A698783-1G A698783-250MG A698783-5G
77117-902EA 29.71 USD
77117-902 77117-904 77117-906 77117-908
2-(Cyclopropylmethoxy)isonicotinonitrile 95%
2-(Cyclopropylmethoxy)isonicotinonitrile

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂O
MDL Number: MFCD11136691
CAS Number: 1019568-08-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR